Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC27
Entrez Gene ID
402783
Gene Name
DnaJ heat shock protein family (Hsp40) member C27
Gene Aliases
RBJ
Chromosome Location
Chr.3: 105433713 - 105439944 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213558.1 NP_998723.1 2 - 3 163 79

View Full Product Details