Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UMOD
Entrez Gene ID
404754
Gene Name
uromodulin
Gene Aliases
ZPD
Chromosome Location
Chr.11: 523817 - 527808 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213576.2 NP_998741.2 5 - 6 637 85
XM_015292282.2 XP_015147768.1 4 - 5 625 85
XM_015292283.2 XP_015147769.1 4 - 5 608 85
GenBank mRNA AB114441.2 - 5 - 6 637 85

View Full Product Details