Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO22
Entrez Gene ID
415355
Gene Name
F-box protein 22
Gene Aliases
-
Chromosome Location
Chr.10: 3943581 - 3950138 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030545.1 NP_001025716.1 6 - 7 800 80
GenBank mRNA AJ719795.1 - 6 - 7 800 80

View Full Product Details