Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THOC3
Entrez Gene ID
416232
Gene Name
THO complex 3
Gene Aliases
-
Chromosome Location
Chr.13: 11143979 - 11147477 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030611.2 NP_001025782.2 4 - 5 709 79
GenBank mRNA AJ720936.1 - 4 - 5 731 79
CR733192.1 - 4 - 5 695 79

View Full Product Details