Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNOT8
Entrez Gene ID
416255
Gene Name
CCR4-NOT transcription complex subunit 8
Gene Aliases
-
Chromosome Location
Chr.13: 12739327 - 12745830 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030618.2 NP_001025789.1 2 - 3 239 99
XM_015293770.2 XP_015149256.1 1 - 2 400 99
XM_015293771.2 XP_015149257.1 2 - 3 268 99
XM_015293772.2 XP_015149258.1 2 - 3 238 99
GenBank mRNA AJ720175.1 - 2 - 3 241 99

View Full Product Details