Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNOT8
Entrez Gene ID
416255
Gene Name
CCR4-NOT transcription complex subunit 8
Gene Aliases
-
Chromosome Location
Chr.13: 12739327 - 12745830 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030618.2 NP_001025789.1 6 - 7 843 96
XM_015293770.2 XP_015149256.1 5 - 6 1004 96
XM_015293771.2 XP_015149257.1 6 - 7 872 96
XM_015293772.2 XP_015149258.1 6 - 7 842 96
GenBank mRNA AJ720175.1 - 6 - 7 845 96

View Full Product Details