Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THRAP3
Entrez Gene ID
419623
Gene Name
thyroid hormone receptor associated protein 3
Gene Aliases
-
Chromosome Location
Chr.23: 4205130 - 4226687 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305665.1 NP_001292594.1 5 - 6 2091 128
XM_015297658.2 XP_015153144.1 7 - 8 2325 128
XM_015297659.1 XP_015153145.1 6 - 7 2116 128
GenBank mRNA AJ719380.1 - 8 - 9 2340 128

View Full Product Details