Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NCDN
Entrez Gene ID
419633
Gene Name
neurochondrin
Gene Aliases
-
Chromosome Location
Chr.23: 4454635 - 4458621 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030901.1 NP_001026072.1 4 - 5 1633 76
GenBank mRNA AJ720824.1 - 4 - 5 1633 76

View Full Product Details