Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDR16C5
Entrez Gene ID
421130
Gene Name
short chain dehydrogenase/reductase family 16C member 5
Gene Aliases
RDHE2
Chromosome Location
Chr.2: 111306862 - 111320568 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031022.1 NP_001026193.1 4 - 5 725 95
GenBank mRNA AJ719902.1 - 4 - 5 725 95

View Full Product Details