Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJB12
Entrez Gene ID
423707
Gene Name
DnaJ heat shock protein family (Hsp40) member B12
Gene Aliases
-
Chromosome Location
Chr.6: 12152738 - 12167831 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031224.1 NP_001026395.1 3 - 4 459 107
XM_015288206.2 XP_015143692.1 3 - 4 107
GenBank mRNA AJ851623.1 - 3 - 4 482 107

View Full Product Details