Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACUL1
Entrez Gene ID
423926
Gene Name
CDK2 associated cullin domain 1
Gene Aliases
C10orf46, C6H10ORF46, CAC1, cullin
Chromosome Location
Chr.6: 30764718 - 30804284 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031238.1 NP_001026409.1 5 - 6 876 74
GenBank mRNA AJ719355.1 - 5 - 6 876 74
CR353948.1 - 3 - 4 446 74

View Full Product Details