Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INSIG2
Entrez Gene ID
424276
Gene Name
insulin induced gene 2
Gene Aliases
INSIG-2
Chromosome Location
Chr.7: 28832351 - 28845172 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305465.1 NP_001292394.1 5 - 6 761 107
NM_001306186.1 NP_001293115.1 5 - 6 601 107
NM_001306187.1 NP_001293116.1 4 - 5 485 107
XM_015289825.2 XP_015145311.1 5 - 6 794 107
GenBank mRNA AJ851538.1 - 5 - 6 575 107

View Full Product Details