Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
INSIG2
Entrez Gene ID
424276
Gene Name
insulin induced gene 2
Gene Aliases
INSIG-2
Chromosome Location
Chr.7: 28832351 - 28845172 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ851538.1 - 6 - 7 2028 145

View Full Product Details