Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB8A
Entrez Gene ID
428352
Gene Name
RAB8A, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.28: 4524689 - 4537499 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031504.1 NP_001026675.1 14 - 15 1855 131
GenBank mRNA AJ851430.1 - 14 - 15 1855 131
BX930100.2 - 7 - 8 539 131

View Full Product Details