Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAB8A
Entrez Gene ID
428352
Gene Name
RAB8A, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.28: 4524689 - 4537499 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031504.1 NP_001026675.1 9 - 10 1511 75
GenBank mRNA AJ851430.1 - 9 - 10 1511 75
BX930100.2 - 2 - 3 195 75
BX931166.1 - 2 - 3 196 75

View Full Product Details