Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEPT5
Entrez Gene ID
416778
Gene Name
septin 5
Gene Aliases
SEP-05, septin-5
Chromosome Location
Chr.15: 1035336 - 1075936 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030654.2 NP_001025825.2 10 - 11 992 61
XM_015275241.2 XP_015130727.1 9 - 10 1180 61
XM_025155434.1 XP_025011202.1 8 - 9 1213 61
XM_025155435.1 XP_025011203.1 9 - 10 4182 61
XM_025155436.1 XP_025011204.1 10 - 11 4751 61
XM_025155437.1 XP_025011205.1 9 - 10 1130 61
GenBank mRNA AJ446747.1 - 2 - 3 195 61
AJ851383.1 - 10 - 11 992 61

View Full Product Details