Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLF2
Entrez Gene ID
418284
Gene Name
myeloid leukemia factor 2
Gene Aliases
-
Chromosome Location
Chr.1: 77831588 - 77838988 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030776.2 NP_001025947.1 2 - 3 206 57
XM_015293367.2 XP_015148853.1 2 - 3 261 57
XM_015293370.2 XP_015148856.1 2 - 3 230 57
XM_015293372.2 XP_015148858.1 1 - 2 86 57
GenBank mRNA AJ851464.1 - 2 - 3 207 57

View Full Product Details