Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL6
Entrez Gene ID
424762
Gene Name
kelch like family member 6
Gene Aliases
-
Chromosome Location
Chr.9: 2728412 - 2746826 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031303.2 NP_001026474.2 2 - 3 520 84
GenBank mRNA AJ450241.1 - 2 - 3 518 84
AJ851379.1 - 2 - 3 441 84

View Full Product Details