Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRSF7L
Entrez Gene ID
426265
Gene Name
serine and arginine rich splicing factor 7-like
Gene Aliases
SFRS7
Chromosome Location
Chr.3: 16639934 - 16647014 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031368.1 NP_001026539.1 2 - 3 282 112
XM_015283521.2 XP_015139007.1 2 - 3 282 112
GenBank mRNA AJ719404.1 - 2 - 3 282 112

View Full Product Details