Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CIR1
Entrez Gene ID
426529
Gene Name
corepressor interacting with RBPJ, 1
Gene Aliases
CIR
Chromosome Location
Chr.7: 16914162 - 16935076 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031386.1 NP_001026557.1 9 - 10 715 179
GenBank mRNA AJ720981.1 - 9 - 10 715 179

View Full Product Details