Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNIH1
Entrez Gene ID
423561
Gene Name
cornichon family AMPA receptor auxiliary protein 1
Gene Aliases
CNI, CNIH
Chromosome Location
Chr.5: 56658853 - 56664637 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033006.1 NP_001028178.1 4 - 5 435 93
GenBank mRNA AB232678.1 - 4 - 5 435 93
BX929843.1 - 4 - 5 437 93
BX931023.1 - 4 - 5 486 93
BX933003.1 - 4 - 5 416 93

View Full Product Details