Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTN1A1
Entrez Gene ID
420061
Gene Name
butyrophilin subfamily 1 member A1
Gene Aliases
TVC
Chromosome Location
Chr.28: 898598 - 904145 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034817.2 NP_001029989.1 3 - 4 512 60
XM_015299709.2 XP_015155195.1 3 - 4 528 60
GenBank mRNA AY847576.1 - 3 - 4 513 60

View Full Product Details