Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CORO1C
Entrez Gene ID
416893
Gene Name
coronin 1C
Gene Aliases
coronin-1C
Chromosome Location
Chr.15: 6830119 - 6868398 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277868.2 NP_001264797.2 3 - 4 718 65
NM_001277869.2 NP_001264798.1 3 - 4 445 65
XM_025155467.1 XP_025011235.1 3 - 4 431 65
XM_025155468.1 XP_025011236.1 3 - 4 482 65
XM_025155469.1 XP_025011237.1 3 - 4 346 65
GenBank mRNA AJ449366.1 - 3 - 4 414 65
AJ453793.1 - 3 - 4 445 65
AJ719514.1 - 3 - 4 409 65
AJ719582.1 - 3 - 4 439 65
AJ719663.1 - 3 - 4 407 65
AJ720924.1 - - 1044 65
BX930410.1 - 3 - 4 718 65
BX932518.2 - 3 - 4 718 65
CN383670.1 - 3 - 4 409 65

View Full Product Details