Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EWSR1
Entrez Gene ID
417022
Gene Name
EWS RNA binding protein 1
Gene Aliases
-
Chromosome Location
Chr.15: 11435907 - 11457069 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039267.2 NP_001034356.2 16 - 17 1998 87
XM_015294853.2 XP_015150339.1 17 - 18 2004 87
XM_015294854.2 XP_015150340.1 17 - 18 2001 87
XM_015294855.2 XP_015150341.1 16 - 17 2001 87
GenBank mRNA AJ719366.1 - 16 - 17 1996 87

View Full Product Details