Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EWSR1
Entrez Gene ID
417022
Gene Name
EWS RNA binding protein 1
Gene Aliases
-
Chromosome Location
Chr.15: 11435907 - 11457069 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039267.2 NP_001034356.2 5 - 6 456 113
XM_015294853.2 XP_015150339.1 6 - 7 462 113
XM_015294854.2 XP_015150340.1 6 - 7 462 113
XM_015294855.2 XP_015150341.1 5 - 6 462 113
XM_015294856.2 XP_015150342.1 5 - 6 462 113
GenBank mRNA AJ719366.1 - 5 - 6 456 113

View Full Product Details