Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SELENOT
Entrez Gene ID
425041
Gene Name
selenoprotein T
Gene Aliases
-
Chromosome Location
Chr.9: 23785354 - 23791069 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006557.4 NP_001006557.3 2 - 3 326 79
GenBank mRNA AJ450766.1 - 2 - 3 329 79
AJ720396.1 - 2 - 3 262 79
BM439905.1 - 2 - 3 286 79
CN224003.1 - 2 - 3 285 79
DR419695.1 - 2 - 3 320 79

View Full Product Details