Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AOX2
Entrez Gene ID
424072
Gene Name
aldehyde oxidase 2
Gene Aliases
-
Chromosome Location
Chr.7: 11233181 - 11281017 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039601.1 NP_001034690.1 1 - 2 108 119
XM_025152179.1 XP_025007947.1 2 - 3 162 119
GenBank mRNA DQ150103.1 - 1 - 2 108 119

View Full Product Details