Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASIC1
Entrez Gene ID
426883
Gene Name
acid sensing ion channel subunit 1
Gene Aliases
ACCN2
Chromosome Location
Chr.33: 5796495 - 5814160 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040467.1 NP_001035557.1 2 - 3 573 64
XM_015300234.2 XP_015155720.1 3 - 4 1047 64
XM_015300235.2 XP_015155721.1 3 - 4 732 64
GenBank mRNA AY956393.1 - 2 - 3 573 64
CR389178.1 - - 810 64

View Full Product Details