Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASIC1
Entrez Gene ID
426883
Gene Name
acid sensing ion channel subunit 1
Gene Aliases
ACCN2
Chromosome Location
Chr.33: 5796495 - 5814160 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040467.1 NP_001035557.1 7 - 8 1205 63
XM_015300232.2 XP_015155718.1 6 - 7 3679 63
XM_015300233.2 XP_015155719.1 6 - 7 2188 63
XM_015300234.2 XP_015155720.1 8 - 9 1679 63
XM_015300235.2 XP_015155721.1 8 - 9 1364 63
GenBank mRNA AY956393.1 - 7 - 8 1205 63

View Full Product Details