Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP1B4
Entrez Gene ID
422365
Gene Name
ATPase Na+/K+ transporting family member beta 4
Gene Aliases
-
Chromosome Location
Chr.4: 16441325 - 16448522 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044651.1 NP_001038116.1 5 - 6 514 100
XM_025149573.1 XP_025005341.1 5 - 6 543 100
GenBank mRNA DQ358914.1 - 5 - 6 514 100

View Full Product Details