Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COLEC11
Entrez Gene ID
428649
Gene Name
collectin subfamily member 11
Gene Aliases
-
Chromosome Location
Chr.3: 93716227 - 93760597 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044665.1 NP_001038130.1 3 - 4 281 117
XM_015284842.2 XP_015140328.1 4 - 5 8992 117
XM_015284843.2 XP_015140329.1 4 - 5 504 117
XM_015284844.2 XP_015140330.1 3 - 4 396 117
XM_025148811.1 XP_025004579.1 3 - 4 195 117
GenBank mRNA DQ129669.1 - 3 - 4 281 117

View Full Product Details