Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DUT
Entrez Gene ID
769338
Gene Name
deoxyuridine triphosphatase
Gene Aliases
-
Chromosome Location
Chr.10: 10589871 - 10600015 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079488.1 NP_001072956.1 5 - 6 455 79
XM_015291930.2 XP_015147416.1 6 - 7 594 79
XM_015291931.2 XP_015147417.1 6 - 7 642 79
XM_025153874.1 XP_025009642.1 6 - 7 536 79
GenBank mRNA AJ720091.1 - 5 - 6 455 79

View Full Product Details