Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM38A
Entrez Gene ID
771253
Gene Name
transmembrane protein 38A
Gene Aliases
-
Chromosome Location
Chr.28: 4384817 - 4391056 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079758.1 NP_001073226.1 5 - 6 709 88
XM_025144285.1 XP_025000053.1 6 - 7 776 88
GenBank mRNA AJ720241.1 - 5 - 6 709 88

View Full Product Details