Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNUPN
Entrez Gene ID
770225
Gene Name
snurportin 1
Gene Aliases
-
Chromosome Location
Chr.10: 3144643 - 3153415 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080878.1 NP_001074347.1 3 - 4 445 79
GenBank mRNA AJ720941.1 - 3 - 4 445 79

View Full Product Details