Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CHMP2A
Entrez Gene ID
776036
Gene Name
charged multivesicular body protein 2A
Gene Aliases
-
Chromosome Location
Chr.31: 5623382 - 5628351 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080892.1 NP_001074361.1 5 - 6 564 114
XM_015273064.2 XP_015128550.1 5 - 6 702 114
XM_015273066.2 XP_015128552.1 5 - 6 752 114
GenBank mRNA AJ721103.1 - 5 - 6 564 114
CR389489.1 - 2 - 3 190 114

View Full Product Details