Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLGN1
Entrez Gene ID
424980
Gene Name
neuroligin 1
Gene Aliases
neuroligin-1
Chromosome Location
Chr.9: 18877811 - 19231069 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081502.1 NP_001074971.1 1 - 2 492 140
XM_015291652.2 XP_015147138.1 2 - 3 1418 140
XM_015291653.2 XP_015147139.1 2 - 3 1464 140
XM_015291654.2 XP_015147140.1 2 - 3 960 140
XM_015291655.2 XP_015147141.1 2 - 3 1420 140
GenBank mRNA EF199811.1 - 1 - 2 492 140
GQ489204.1 - 1 - 2 492 140
GU080337.1 - 1 - 2 492 140
GU080338.1 - 1 - 2 492 140
GU574580.1 - 1 - 2 492 140

View Full Product Details