Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLGN1
Entrez Gene ID
424980
Gene Name
neuroligin 1
Gene Aliases
neuroligin-1
Chromosome Location
Chr.9: 18877811 - 19231069 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081502.1 NP_001074971.1 3 - 4 610 115
XM_015291652.2 XP_015147138.1 4 - 5 1536 115
XM_015291653.2 XP_015147139.1 4 - 5 1582 115
XM_015291654.2 XP_015147140.1 4 - 5 1078 115
XM_015291655.2 XP_015147141.1 4 - 5 1538 115
XM_015291656.2 XP_015147142.1 3 - 4 1475 115
XM_015291658.2 XP_015147144.1 3 - 4 1476 115
GenBank mRNA BX935583.2 - 3 - 4 1109 115
EF199811.1 - 3 - 4 610 115
GQ489204.1 - 3 - 4 610 115
GU080337.1 - 3 - 4 610 115
GU080339.1 - 2 - 3 550 115
GU574580.1 - 3 - 4 610 115

View Full Product Details