The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098608.1 | NP_001092078.1 | 3 - 4 | 330 | 82 |
XM_015291403.2 | XP_015146889.1 | 3 - 4 | 379 | 82 | |
XM_025153364.1 | XP_025009132.1 | 3 - 4 | 435 | 82 | |
XM_025153365.1 | XP_025009133.1 | 3 - 4 | 431 | 82 | |
XM_025153366.1 | XP_025009134.1 | 3 - 4 | 462 | 82 | |
XM_025153367.1 | XP_025009135.1 | 3 - 4 | 462 | 82 | |
XM_025153368.1 | XP_025009136.1 | 3 - 4 | 384 | 82 | |
GenBank mRNA | CR352395.1 | - | 3 - 4 | 361 | 82 |
CR389564.1 | - | 3 - 4 | 356 | 82 | |
EF551041.1 | - | 3 - 4 | 330 | 82 | |
EU334507.1 | - | 2 - 3 | 324 | 82 |