Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL4I1
Entrez Gene ID
417039
Gene Name
interleukin 4 induced 1
Gene Aliases
LAO
Chromosome Location
Chr.16: 2473441 - 2478234 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099351.3 NP_001092821.2 4 - 5 609 62

View Full Product Details