Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGMAT
Entrez Gene ID
373942
Gene Name
agmatinase
Gene Aliases
AUH
Chromosome Location
Chr.21: 4911063 - 4915112 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204143.3 NP_989474.1 3 - 4 577 93
XM_025142476.1 XP_024998244.1 4 - 5 2174 93
GenBank mRNA AF401291.2 - 3 - 4 577 93

View Full Product Details