Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FABP7
Entrez Gene ID
396246
Gene Name
fatty acid binding protein 7
Gene Aliases
-
Chromosome Location
Chr.3: 61282801 - 61286154 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205308.2 NP_990639.1 2 - 3 258 82
GenBank mRNA X65459.1 - - 258 82

View Full Product Details