Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RLN3
Entrez Gene ID
427223
Gene Name
relaxin 3
Gene Aliases
RFLB, RLN1, RLX3
Chromosome Location
Chr.Z: 27625033 - 27627531 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113200.1 NP_001106671.1 1 - 2 331 73
GenBank mRNA BU138425.1 - 1 - 2 334 73
EU442364.1 - 1 - 2 228 73

View Full Product Details