Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOG
Entrez Gene ID
373912
Gene Name
noggin
Gene Aliases
-
Chromosome Location
Chr.18: 6188544 - 6189215 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204123.1 NP_989454.1 1 - 1 551 62
GenBank mRNA AB013493.1 - 1 - 1 392 62
AF057364.1 - 1 - 1 551 62

View Full Product Details