Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POU1F1
Entrez Gene ID
374215
Gene Name
POU class 1 homeobox 1
Gene Aliases
GHF-1, PIT-1, PIT1, Pit-1*
Chromosome Location
Chr.1: 94255838 - 94273551 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204319.1 NP_989650.1 4 - 5 793 67
XM_015295651.2 XP_015151137.1 5 - 6 2934 67
XM_015295656.2 XP_015151142.1 5 - 6 2850 67
GenBank mRNA AF029892.1 - 4 - 5 793 67
AJ222692.2 - 4 - 5 709 67
AJ236855.1 - 5 - 6 733 67
AJ236856.1 - 5 - 6 817 67

View Full Product Details