Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPVF
Entrez Gene ID
378785
Gene Name
neuropeptide VF precursor
Gene Aliases
RFRP
Chromosome Location
Chr.2: 31800647 - 31804213 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204363.1 NP_989694.1 1 - 2 179 107
GenBank mRNA AB120325.1 - 1 - 2 179 107
AY442186.1 - 1 - 2 141 107
CR406155.1 - 1 - 2 156 107

View Full Product Details