Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS4
Entrez Gene ID
378901
Gene Name
regulator of G-protein signaling 4
Gene Aliases
-
Chromosome Location
Chr.8: 3972117 - 4072191 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204385.1 NP_989716.1 1 - 2 144 56
XM_025152734.1 XP_025008502.1 1 - 2 134 56
GenBank mRNA AY297457.1 - 1 - 2 144 56
BX950639.2 - 1 - 2 144 56

View Full Product Details