Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FUT10
Entrez Gene ID
395073
Gene Name
fucosyltransferase 10
Gene Aliases
-
Chromosome Location
Chr.Z: 53942718 - 53953349 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204430.2 NP_989761.2 3 - 4 577 93
XM_015280410.2 XP_015135896.1 3 - 4 664 93
GenBank mRNA AJ535692.1 - 3 - 4 579 93
BX929517.2 - 2 - 3 401 93

View Full Product Details