Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNAI1
Entrez Gene ID
396047
Gene Name
snail family transcriptional repressor 1
Gene Aliases
-
Chromosome Location
Chr.20: 13790862 - 13793593 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205142.1 NP_990473.1 1 - 2 98 70
GenBank mRNA Y09905.1 - - 98 70

View Full Product Details