Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP20
Entrez Gene ID
415627
Gene Name
cilia and flagella associated protein 20
Gene Aliases
C11H16ORF80, C16orf80, GTL3
Chromosome Location
Chr.11: 431116 - 437630 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005833.1 NP_001005833.1 5 - 6 680 75
GenBank mRNA AJ721091.1 - 5 - 6 680 75
CR386276.1 - 5 - 6 687 75

View Full Product Details