Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL9
Entrez Gene ID
416623
Gene Name
methyltransferase like 9
Gene Aliases
-
Chromosome Location
Chr.14: 9132062 - 9150363 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006172.2 NP_001006172.1 1 - 2 290 143
XM_015294230.2 XP_015149716.1 1 - 2 284 143
GenBank mRNA AJ719408.1 - 1 - 2 292 143

View Full Product Details